siRNA Details
| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020717.3 | NP_065768.2 | 4 | 566 | ||
| NR_027121.1 | 4 | 566 | ||||
| XM_017029682.1 | XP_016885171.1 | 4 | 672 | |||
| XM_017029683.1 | XP_016885172.1 | 3 | 537 | |||
| XM_017029684.1 | XP_016885173.1 | 3 | 689 | |||
| XM_017029685.1 | XP_016885174.1 | 4 | 689 | |||
| XM_017029686.1 | XP_016885175.1 | 2 | 554 | |||
| XM_017029687.1 | XP_016885176.1 | 4 | 676 | |||
| GenBank mRNA | AB033028.1 | - | 4 | 566 | ||
| AY044234.1 | - | 3 | 647 | |||
| BC151240.1 | - | 4 | 577 | 8860513 | ||

SHROOM4
Genomic Map