siRNA Details
| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020717.3 | NP_065768.2 | 9 | 4390 | ||
| NR_027121.1 | 9 | 4390 | ||||
| XM_017029682.1 | XP_016885171.1 | 10 | 4610 | |||
| XM_017029683.1 | XP_016885172.1 | 9 | 4475 | |||
| XM_017029684.1 | XP_016885173.1 | 9 | 4627 | |||
| XM_017029686.1 | XP_016885175.1 | 8 | 4492 | |||
| GenBank mRNA | AB033028.1 | - | 9 | 4405 | ||
| BC151240.1 | - | 9 | 4416 | 8860513 | ||

SHROOM4
Genomic Map