siRNA Details
| Gene Symbol: USP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020718.3 | NP_065769.3 | 6 | 1150 | ||
| XM_017023496.1 | XP_016878985.1 | 6 | 1181 | |||
| XM_017023497.1 | XP_016878986.1 | 6 | 2676 | |||
| XM_017023498.1 | XP_016878987.1 | 5 | 463 | |||
| XM_017023499.1 | XP_016878988.1 | 3 | 267 | |||
| XM_017023500.1 | XP_016878989.1 | 6 | 1181 | |||
| GenBank mRNA | AJ586135.2 | - | 6 | 1150 | ||
| AK126752.1 | - | 6 | 834 | |||

USP31
Genomic Map