siRNA Details
| Gene Symbol: KIAA1210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020721.1 | NP_065772.1 | 12 | 4731 | ||
| XM_006724774.3 | XP_006724837.1 | 10 | 4320 | |||
| XM_017029688.1 | XP_016885177.1 | 10 | 4359 | |||
| XM_017029689.1 | XP_016885178.1 | 9 | 4161 | |||
| GenBank mRNA | AB033036.1 | - | 2 | 2950 | ||

KIAA1210
Genomic Map