siRNA Details
| Gene Symbol: NLGN4X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282145.1 | NP_001269074.1 | 7 | 2731 | ||
| NM_001282146.1 | NP_001269075.1 | 6 | 2406 | |||
| NM_020742.3 | NP_065793.1 | 6 | 2241 | |||
| NM_181332.2 | NP_851849.1 | 6 | 2424 | |||
| XM_005274564.2 | XP_005274621.1 | 7 | 2478 | |||
| XM_005274565.2 | XP_005274622.1 | 7 | 2461 | |||
| XM_005274566.4 | XP_005274623.1 | 7 | 2405 | |||
| XM_006724504.3 | XP_006724567.1 | 7 | 2346 | |||
| XM_011545547.2 | XP_011543849.1 | 7 | 2480 | |||
| XM_011545548.2 | XP_011543850.1 | 7 | 2644 | |||
| XM_017029690.1 | XP_016885179.1 | 7 | 2290 | |||
| XM_017029691.1 | XP_016885180.1 | 6 | 2420 | |||
| XM_017029692.1 | XP_016885181.1 | 6 | 2345 | |||
| XM_017029693.1 | XP_016885182.1 | 6 | 2230 | |||
| GenBank mRNA | AB033086.1 | - | 6 | 2241 | ||
| AF376803.1 | - | 6 | 2241 | |||
| AK022621.1 | - | 3 | 1075 | |||
| AK055471.1 | - | 7 | 2464 | |||
| AK291036.1 | - | 6 | 2278 | |||
| AX773938.1 | - | 7 | 2352 | |||
| AY358562.1 | - | 6 | 1826 | |||
| BC034018.1 | - | 6 | 2207 | 4702963 | ||

NLGN4X
Genomic Map