siRNA Details
| Gene Symbol: INTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020748.2 | NP_065799.1 | 25 | 3617 | ||
| NR_026641.1 | 25 | 3898 | ||||
| XM_006721987.3 | XP_006722050.1 | 25 | 3624 | |||
| XM_011525058.2 | XP_011523360.1 | 25 | 3904 | |||
| GenBank mRNA | AB033113.1 | - | 25 | 3845 | ||
| AK299032.1 | - | 25 | 3873 | |||
| AK308307.1 | - | 25 | 3582 | |||
| AL136800.1 | - | 25 | 3617 | |||
| CR533582.1 | - | 24 | 3541 | |||

INTS2
Genomic Map