siRNA Details
| Gene Symbol: INTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020748.2 | NP_065799.1 | 17 | 2308 | ||
| NR_026641.1 | 17 | 2589 | ||||
| XM_006721987.3 | XP_006722050.1 | 17 | 2315 | |||
| XM_011525058.2 | XP_011523360.1 | 17 | 2595 | |||
| GenBank mRNA | AB033113.1 | - | 17 | 2536 | ||
| AK299032.1 | - | 17 | 2564 | |||
| AK308307.1 | - | 17 | 2273 | |||
| AL136800.1 | - | 17 | 2308 | |||
| CR533582.1 | - | 16 | 2232 | |||

INTS2
Genomic Map