siRNA Details
| Gene Symbol: GPR158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020752.2 | NP_065803.2 | 4 | 1291 | ||
| GenBank mRNA | AK095120.1 | - | 4 | 1589 | ||
| AY528411.1 | - | 4 | 1291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR158| Gene Symbol: GPR158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020752.2 | NP_065803.2 | 4 | 1291 | ||
| GenBank mRNA | AK095120.1 | - | 4 | 1589 | ||
| AY528411.1 | - | 4 | 1291 | |||