siRNA Details
| Gene Symbol: RGAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020769.2 | NP_065820.1 | 3 | 483 | ||
| XM_011530998.2 | XP_011529300.1 | 3 | 505 | |||
| XM_011530999.2 | XP_011529301.1 | 4 | 1354 | |||
| XM_017029694.1 | XP_016885183.1 | 2 | 413 | |||
| XM_017029695.1 | XP_016885184.1 | 5 | 1487 | |||
| GenBank mRNA | AB037739.1 | - | 3 | 483 | ||
| AK302427.1 | - | 3 | 505 | |||
| AY121804.1 | - | 3 | 483 | |||
| BC113670.1 | - | 2 | 324 | 8322722 | ||
| BC117184.1 | - | 2 | 324 | 40125735 | ||

RGAG1
Genomic Map