siRNA Details
| Gene Symbol: KIAA1328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322327.1 | NP_001309256.1 | 2 | 108 | ||
| NM_020776.1 | NP_065827.1 | 2 | 97 | |||
| NR_136303.1 | 2 | 108 | ||||
| NR_136304.1 | 2 | 108 | ||||
| NR_136305.1 | 2 | 108 | ||||
| XM_005258315.4 | XP_005258372.1 | 2 | 100 | |||
| XM_005258317.3 | XP_005258374.1 | 2 | 489 | |||
| XM_005258318.4 | XP_005258375.1 | 2 | 104 | |||
| XM_011526099.2 | XP_011524401.1 | 2 | 293 | |||
| XM_011526100.2 | XP_011524402.1 | 2 | 100 | |||
| XM_017025876.1 | XP_016881365.1 | 2 | 410 | |||
| XM_017025877.1 | XP_016881366.1 | 2 | 36 | |||
| XM_017025878.1 | XP_016881367.1 | 2 | 285 | |||
| GenBank mRNA | AB037749.2 | - | 2 | 489 | ||
| AK292288.1 | - | 2 | 86 | |||
| AK304230.1 | - | 2 | 108 | |||
| AL832625.2 | - | 2 | 98 | |||
| BX648875.1 | - | 2 | 84 | |||

KIAA1328
Genomic Map