siRNA Details
| Gene Symbol: ALPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020778.4 | NP_065829.3 | 5 | 2203 | ||
| XM_017022444.1 | XP_016877933.1 | 4 | 2201 | |||
| XM_017022445.1 | XP_016877934.1 | 5 | 2252 | |||
| GenBank mRNA | AL833288.1 | - | 1 | 885 | ||
| AY044449.1 | - | 5 | 2203 | |||

ALPK3
Genomic Map