siRNA Details
| Gene Symbol: ALPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020778.4 | NP_065829.3 | 6 | 2541 | ||
| XM_017022444.1 | XP_016877933.1 | 5 | 2539 | |||
| XM_017022445.1 | XP_016877934.1 | 6 | 2590 | |||
| GenBank mRNA | AL833288.1 | - | 2 | 1223 | ||
| AY044449.1 | - | 6 | 2541 | |||

ALPK3
Genomic Map