siRNA Details
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 25 | 3037 | ||
NM_020779.3 | NP_065830.2 | 24 | 3004 | |||
XM_011533007.2 | XP_011531309.1 | 14 | 1729 | |||
GenBank mRNA | AB037757.2 | - | 25 | 3045 | ||
AK091205.1 | - | 16 | 2096 | |||
AK122917.1 | - | 24 | 2929 | |||
AK315094.1 | - | 24 | 2833 | |||
BC036659.2 | - | 24 | 3003 | 5266940 |
