siRNA Details
| Gene Symbol: DISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020780.1 | NP_065831.1 | 12 | 2632 | ||
| XM_011541828.2 | XP_011540130.1 | 11 | 4734 | |||
| XM_011541829.1 | XP_011540131.1 | 11 | 1607 | |||
| XM_011541830.2 | XP_011540132.1 | 12 | 2715 | |||
| XM_011541831.2 | XP_011540133.1 | 12 | 2715 | |||
| XM_011541832.2 | XP_011540134.1 | 12 | 2715 | |||
| XM_011541833.2 | XP_011540135.1 | 12 | 2715 | |||
| GenBank mRNA | AB037758.1 | - | 12 | 2632 | ||
| AB593117.1 | - | 12 | 2715 | |||
| AL117236.1 | - | 11 | 1552 | |||

DISP3
Genomic Map