siRNA Details
| Gene Symbol: ZNF398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020781.3 | NP_065832.1 | 4, 5 | 886 | ||
| NM_170686.2 | NP_733787.1 | 3, 4 | 823 | |||
| XM_011516439.2 | XP_011514741.1 | 4, 5 | 704 | |||
| XM_011516440.2 | XP_011514742.1 | 3, 4 | 676 | |||
| XM_011516441.2 | XP_011514743.1 | 3, 4 | 659 | |||
| GenBank mRNA | AK290499.1 | - | 3, 4 | 791 | ||
| AK301890.1 | - | 4, 5 | 704 | |||
| AK304603.1 | - | 1, 2 | 122 | |||
| AY049743.1 | - | 4, 5 | 886 | |||
| AY049744.1 | - | 3, 4 | 565 | |||
| BC043295.2 | - | 3, 4 | 815 | 5298104 | ||

ZNF398
Genomic Map