siRNA Details
| Gene Symbol: KLHL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020782.1 | NP_065833.1 | 2, 3 | 1146 | ||
| GenBank mRNA | AB037761.1 | - | 2, 3 | 877 | ||
| AK095405.1 | - | 2, 3 | 1146 | |||
| BC028742.1 | - | 3, 4 | 1014 | |||
| BC108669.1 | - | 2, 3 | 1117 | 6173072 | ||
| BC137116.1 | - | 2, 3 | 1112 | 9021113 | ||
| BC137117.1 | - | 2, 3 | 1112 | 9021114 | ||

KLHL42
Genomic Map