siRNA Details
| Gene Symbol: KLHL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020782.1 | NP_065833.1 | 3 | 1513 | ||
| XM_017019698.1 | XP_016875187.1 | 3 | 2278 | |||
| GenBank mRNA | AB037761.1 | - | 3 | 1244 | ||
| AK095405.1 | - | 3 | 1513 | |||
| BC028742.1 | - | 4 | 1381 | |||
| BC108669.1 | - | 3 | 1484 | 6173072 | ||
| BC137116.1 | - | 3 | 1479 | 9021113 | ||
| BC137117.1 | - | 3 | 1479 | 9021114 | ||

KLHL42
Genomic Map