siRNA Details
| Gene Symbol: ZNF624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020787.3 | NP_065838.2 | 6 | 1623 | ||
| XM_006721562.3 | XP_006721625.1 | 6 | 1804 | |||
| XM_011523970.2 | XP_011522272.1 | 6 | 2974 | |||
| GenBank mRNA | AB037770.1 | - | 1 | 1444 | ||
| AK131401.1 | - | 6 | 1614 | |||
| BC103943.1 | - | 1 | 1205 | 40008882 | ||
| BC103944.1 | - | 1 | 1205 | 40008883 | ||
| BC103946.1 | - | 1 | 1205 | 40008885 | ||

ZNF624
Genomic Map