siRNA Details
| Gene Symbol: IGSF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135050.1 | NP_001128522.1 | 14 | 1948 | ||
| NM_020789.3 | NP_065840.2 | 14 | 1900 | |||
| XM_017001884.1 | XP_016857373.1 | 14 | 2013 | |||
| GenBank mRNA | AB037776.1 | - | 14 | 1948 | ||
| AK056650.1 | - | 4 | 1777 | |||
| AY203932.1 | - | 3 | 1253 | |||
| BC030141.2 | - | 14 | 1900 | 4762143 | ||

IGSF9
Genomic Map