siRNA Details
| Gene Symbol: USP35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020798.2 | NP_065849.1 | 10 | 3234 | ||
| XM_011545187.2 | XP_011543489.1 | 10 | 2377 | |||
| XM_011545188.2 | XP_011543490.1 | 8 | 2160 | |||
| XM_017018052.1 | XP_016873541.1 | 10 | 3231 | |||
| XM_017018053.1 | XP_016873542.1 | 9 | 2226 | |||
| GenBank mRNA | AB037793.1 | - | 7 | 2697 | ||
| AJ586137.1 | - | 9 | 2875 | |||
| AK056231.1 | - | 5 | 2027 | |||
| AK301048.1 | - | 2 | 1625 | |||
| BC131489.1 | - | 8 | 2201 | 40038029 | ||

USP35
Genomic Map