siRNA Details
| Gene Symbol: KLHL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020805.2 | NP_065856.1 | 3 | 1431 | ||
| XM_011526109.2 | XP_011524411.2 | 3 | 1123 | |||
| GenBank mRNA | AB037805.1 | - | 3 | 1431 | ||
| AK294996.1 | - | 2 | 669 | |||
| BC021267.2 | - | 3 | 1299 | 4588702 | ||

KLHL14
Genomic Map