siRNA Details
| Gene Symbol: KLHL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020805.2 | NP_065856.1 | 7 | 1881 | ||
| GenBank mRNA | AB037805.1 | - | 7 | 1881 | ||
| AK294996.1 | - | 6 | 1119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHL14| Gene Symbol: KLHL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020805.2 | NP_065856.1 | 7 | 1881 | ||
| GenBank mRNA | AB037805.1 | - | 7 | 1881 | ||
| AK294996.1 | - | 6 | 1119 | |||