siRNA Details
| Gene Symbol: TMEM181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020823.1 | NP_065874.1 | 10, 11 | 1320 | ||
| XM_005267074.3 | XP_005267131.1 | 10, 11 | 1037 | |||
| XM_011535999.1 | XP_011534301.1 | 10, 11 | 1348 | |||
| XM_011536000.1 | XP_011534302.1 | 10, 11 | 1348 | |||
| XM_011536001.2 | XP_011534303.1 | 10, 11 | 1528 | |||
| XM_011536002.2 | XP_011534304.1 | 10, 11 | 1502 | |||
| GenBank mRNA | AB037844.1 | - | 10, 11 | 1322 | ||
| BC033450.1 | - | 9, 10 | 1043 | |||

TMEM181
Genomic Map