siRNA Details
| Gene Symbol: CRAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020825.3 | NP_065876.3 | 5 | 791 | ||
| GenBank mRNA | AB037847.2 | - | 3 | 184 | ||
| BC137172.1 | - | 4 | 341 | 9021169 | ||
| BC137173.1 | - | 4 | 341 | 9021170 | ||
| BC152477.1 | - | 3 | 184 | 8862615 | ||
| BE048705.1 | - | 5 | 531 | |||
| BM451163.1 | - | 2 | 89 | |||

CRAMP1
Genomic Map