siRNA Details
| Gene Symbol: WDFY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020830.4 | NP_065881.1 | 8, 9 | 943 | ||
| GenBank mRNA | AB037856.1 | - | 8, 9 | 856 | ||
| AF411977.1 | - | 8, 9 | 840 | |||
| AJ310568.1 | - | 8, 9 | 930 | |||
| AK022888.1 | - | 8, 9 | 869 | |||
| AK023415.1 | - | 7, 8 | 737 | |||
| BC040525.1 | - | 8, 9 | 855 | 5285567 | ||
| BC065934.1 | - | 8, 9 | 887 | 6164567 | ||

WDFY1
Genomic Map