siRNA Details
| Gene Symbol: HOMEZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020834.2 | NP_065885.2 | 2 | 639 | ||
| GenBank mRNA | AB037864.1 | - | 2 | 565 | ||
| AF463523.1 | - | 2 | 563 | |||
| AK296927.1 | - | 2 | 599 | |||
| AK301849.1 | - | 2 | 485 | |||
| AK302788.1 | - | 2 | 499 | |||
| BC130392.1 | - | 1 | 464 | 40146423 | ||

HOMEZ
Genomic Map