siRNA Details
| Gene Symbol: BAHCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291324.1 | NP_001278253.1 | 5, 6 | 2352 | ||
| XM_011525063.2 | XP_011523365.1 | 6, 7 | 2744 | |||
| XM_017024898.1 | XP_016880387.1 | 5, 6 | 2403 | |||
| XM_017024899.1 | XP_016880388.1 | 5, 6 | 2402 | |||
| GenBank mRNA | AB040880.1 | - | 1, 2 | 103 | ||

BAHCC1
Genomic Map