siRNA Details
| Gene Symbol: WDR48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303402.1 | NP_001290331.1 | 4 | 401 | ||
| NM_001303403.1 | NP_001290332.1 | 5 | 453 | |||
| NM_020839.3 | NP_065890.1 | 5 | 480 | |||
| XM_005265346.3 | XP_005265403.1 | 5 | 480 | |||
| XM_011533971.1 | XP_011532273.1 | 5 | 592 | |||
| XM_017006960.1 | XP_016862449.1 | 5 | 592 | |||
| GenBank mRNA | AB040882.2 | - | 5 | 451 | ||
| AF468833.1 | - | 5 | 440 | |||
| AK025513.1 | - | 5 | 450 | |||
| AK297349.1 | - | 4 | 371 | |||
| AK302307.1 | - | 5 | 423 | |||
| AL832926.1 | - | 5 | 434 | |||
| BC026353.1 | - | 5 | 450 | 4812745 | ||
| DA215447.1 | - | 5 | 480 | |||

WDR48
Genomic Map