siRNA Details
| Gene Symbol: SLAIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020846.1 | NP_065897.1 | 5 | 1103 | ||
| XM_005248121.2 | XP_005248178.1 | 5 | 1219 | |||
| GenBank mRNA | AB040891.1 | - | 5 | 1073 | ||
| AK025264.1 | - | 1 | 154 | |||
| AK026476.1 | - | 5 | 650 | |||
| AK291006.1 | - | 5 | 1233 | |||
| BC031691.2 | - | 3 | 423 | |||
| BC040993.1 | - | 5 | 1382 | 5272534 | ||

SLAIN2
Genomic Map