siRNA Details
| Gene Symbol: RANBP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320238.1 | NP_001307167.1 | 9 | 1340 | ||
| NM_001320239.1 | NP_001307168.1 | 9 | 1284 | |||
| NM_001320240.1 | NP_001307169.1 | 13 | 1905 | |||
| NM_020850.2 | NP_065901.1 | 10 | 1452 | |||
| XM_005256070.2 | XP_005256127.1 | 10 | 1360 | |||
| XM_005256072.2 | XP_005256129.1 | 9 | 1307 | |||
| XM_006721238.3 | XP_006721301.1 | 5 | 1117 | |||
| XM_011523245.2 | XP_011521547.1 | 7 | 1099 | |||
| XM_011523247.2 | XP_011521549.1 | 7 | 905 | |||
| XM_017023507.1 | XP_016878996.1 | 7 | 966 | |||
| XM_017023508.1 | XP_016878997.1 | 11 | 1520 | |||
| XM_017023509.1 | XP_016878998.1 | 8 | 955 | |||
| GenBank mRNA | AB040897.1 | - | 10 | 1341 | ||
| AK000686.1 | - | 1583 | ||||
| AK055727.1 | - | 13 | 1810 | |||
| AK295530.1 | - | 9 | 1248 | |||
| AK298806.1 | - | 9 | 1211 | |||
| AY337313.1 | - | 10 | 1452 | |||
| BC099917.1 | - | 10 | 1357 | 30528481 | ||
| BC121176.1 | - | 10 | 1450 | 40122561 | ||
| BC121177.2 | - | 10 | 1450 | 40122562 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_145824, AK173177, BC024698, EU281316, AY337314.

RANBP10
Genomic Map