siRNA Details
| Gene Symbol: LRFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020862.1 | NP_065913.1 | 2 | 1435 | ||
| XM_005259102.4 | XP_005259159.1 | 5 | 1635 | |||
| XM_017027033.1 | XP_016882522.1 | 5 | 1750 | |||
| GenBank mRNA | AB040917.1 | - | 2 | 1224 | ||
| BC014678.1 | - | 531 | ||||
| BC025310.1 | - | 2 | 748 | |||

LRFN1
Genomic Map