siRNA Details
| Gene Symbol: ZSWIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020883.1 | NP_065934.1 | 14 | 2983 | ||
| XM_017001913.1 | XP_016857402.1 | 12 | 1892 | |||
| GenBank mRNA | AB040944.1 | - | 14 | 2961 | ||
| BC013836.1 | - | 8 | 1134 | |||

ZSWIM5
Genomic Map