siRNA Details
Gene Symbol: ZSWIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020883.1 | NP_065934.1 | 3 | 1217 | ||
XM_011541861.2 | XP_011540163.1 | 3 | 1225 | |||
XM_017001913.1 | XP_016857402.1 | 2 | 303 | |||
GenBank mRNA | AB040944.1 | - | 3 | 1195 |
