siRNA Details
| Gene Symbol: KIAA1522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198972.1 | NP_001185901.1 | 6 | 1019 | ||
| NM_020888.2 | NP_065939.2 | 6 | 1104 | |||
| XM_006710791.3 | XP_006710854.1 | 6 | 1038 | |||
| XM_017001916.1 | XP_016857405.1 | 4 | 702 | |||
| XM_017001917.1 | XP_016857406.1 | 7 | 1066 | |||
| GenBank mRNA | AB040955.1 | - | 6 | 986 | ||

KIAA1522
Genomic Map