siRNA Details
| Gene Symbol: ZBTB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128833.1 | NP_001122305.1 | 4 | 3164 | ||
| NM_020899.3 | NP_065950.2 | 4 | 3240 | |||
| XM_006721563.3 | XP_006721626.1 | 4 | 3091 | |||
| XM_006721564.2 | XP_006721627.1 | 4 | 3071 | |||
| XM_011523972.2 | XP_011522274.1 | 4 | 3523 | |||
| GenBank mRNA | AB040971.2 | - | 4 | 3164 | ||
| AK027143.1 | - | 1 | 315 | |||
| AK122826.1 | - | 2 | 2492 | |||
| AK122971.1 | - | 4 | 3064 | |||
| AY302699.1 | - | 4 | 3080 | |||
| BC033259.1 | - | 1 | 786 | |||
| BC043352.2 | - | 4 | 3189 | 6175382 | ||

ZBTB4
Genomic Map