siRNA Details
| Gene Symbol: ZBTB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128833.1 | NP_001122305.1 | 4 | 1769 | ||
| NM_020899.3 | NP_065950.2 | 4 | 1845 | |||
| XM_006721563.3 | XP_006721626.1 | 4 | 1696 | |||
| XM_006721564.2 | XP_006721627.1 | 4 | 1676 | |||
| XM_011523972.2 | XP_011522274.1 | 4 | 2128 | |||
| GenBank mRNA | AB040971.2 | - | 4 | 1769 | ||
| AK122826.1 | - | 2 | 1097 | |||
| AK122971.1 | - | 4 | 1669 | |||
| AY302699.1 | - | 4 | 1685 | |||
| BC043352.2 | - | 4 | 1794 | 6175382 | ||

ZBTB4
Genomic Map