siRNA Details
| Gene Symbol: PLEKHA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161354.1 | NP_001154826.1 | 3 | 651 | ||
| NM_020904.2 | NP_065955.2 | 3 | 651 | |||
| XM_005259107.2 | XP_005259164.1 | 3 | 646 | |||
| XM_006723301.2 | XP_006723364.1 | 3 | 646 | |||
| XM_011527157.1 | XP_011525459.1 | 3 | 646 | |||
| XM_011527159.1 | XP_011525461.1 | 3 | 646 | |||
| XM_011527160.1 | XP_011525462.1 | 3 | 646 | |||
| GenBank mRNA | AY007233.1 | - | 3 | 625 | ||
| BC024157.2 | - | 3 | 631 | 3953478 | ||

PLEKHA4
Genomic Map