siRNA Details
| Gene Symbol: CACHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293274.1 | NP_001280203.1 | 20, 21 | 3199 | ||
| NM_020925.2 | NP_065976.2 | 21, 22 | 2920 | |||
| XM_011541862.1 | XP_011540164.1 | 20, 21 | 3195 | |||
| XM_011541863.1 | XP_011540165.1 | 21, 22 | 3344 | |||
| GenBank mRNA | AB046793.1 | - | 19, 20 | 2702 | ||
| AK022571.1 | - | 8, 9 | 1018 | |||
| AK024256.1 | - | 11, 12 | 1345 | |||
| AK299361.1 | - | 7, 8 | 763 | |||
| AL833000.1 | - | 11, 12 | 1369 | |||
| BC039301.1 | - | 21, 22 | 2906 | 4836806 | ||
| BX537603.1 | - | 20, 21 | 3198 | |||

CACHD1
Genomic Map