siRNA Details
| Gene Symbol: CACHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293274.1 | NP_001280203.1 | 3 | 698 | ||
| NM_020925.2 | NP_065976.2 | 3 | 274 | |||
| XM_011541863.1 | XP_011540165.1 | 3 | 698 | |||
| GenBank mRNA | AB046793.1 | - | 1 | 56 | ||
| BC039301.1 | - | 2 | 229 | 4836806 | ||
| BX537603.1 | - | 3 | 697 | |||
| CX163220.1 | - | 3 | 265 | |||

CACHD1
Genomic Map