siRNA Details
| Gene Symbol: KIAA1586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286274.1 | NP_001273203.1 | 3 | 1436 | ||
| NM_001286275.1 | NP_001273204.1 | 4 | 1495 | |||
| NM_001286276.1 | NP_001273205.1 | 4 | 1447 | |||
| NM_020931.3 | NP_065982.1 | 4 | 1517 | |||
| GenBank mRNA | AB046806.1 | - | 1 | 2629 | ||
| AK290988.1 | - | 4 | 1311 | |||
| AK295519.1 | - | 3 | 1242 | |||
| BC029372.1 | - | 4 | 1326 | |||
| BC041125.1 | - | 4 | 1463 | 5785266 | ||

KIAA1586
Genomic Map