siRNA Details
| Gene Symbol: MAGEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020932.2 | NP_065983.1 | 1 | 2578 | ||
| GenBank mRNA | AB046807.1 | - | 1 | 2507 | ||
| AF490507.1 | - | 1 | 2507 | |||
| AK291270.1 | - | 1 | 2493 | |||
| BC050588.1 | - | 1 | 2459 | 6189864 | ||

MAGEE1
Genomic Map