siRNA Details
| Gene Symbol: FAM160B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135051.1 | NP_001128523.1 | 8 | 1360 | ||
| NM_020940.3 | NP_065991.3 | 8 | 1360 | |||
| GenBank mRNA | AB046820.2 | - | 7 | 955 | ||
| BC037207.1 | - | 8 | 1360 | 4828347 | ||
| CR933692.1 | - | 8 | 1080 | |||

FAM160B1
Genomic Map