siRNA Details
| Gene Symbol: KIAA1614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020950.1 | NP_066001.1 | 2 | 744 | ||
| GenBank mRNA | AB046834.1 | - | 2 | 732 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA1614| Gene Symbol: KIAA1614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020950.1 | NP_066001.1 | 2 | 744 | ||
| GenBank mRNA | AB046834.1 | - | 2 | 732 | ||