siRNA Details
| Gene Symbol: ANO8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020959.2 | NP_066010.1 | 11, 12 | 1486 | ||
| XM_017027048.1 | XP_016882537.1 | 11, 12 | 1491 | |||
| GenBank mRNA | AB046843.2 | - | 11, 12 | 1486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANO8| Gene Symbol: ANO8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020959.2 | NP_066010.1 | 11, 12 | 1486 | ||
| XM_017027048.1 | XP_016882537.1 | 11, 12 | 1491 | |||
| GenBank mRNA | AB046843.2 | - | 11, 12 | 1486 | ||