siRNA Details
| Gene Symbol: GPR107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136557.1 | NP_001130029.1 | 1, 2 | 376 | ||
| NM_001136558.1 | NP_001130030.1 | 1, 2 | 376 | |||
| NM_001287346.1 | NP_001274275.1 | 1, 2 | 376 | |||
| NM_020960.4 | NP_066011.2 | 1, 2 | 376 | |||
| XM_017014953.1 | XP_016870442.1 | 1, 2 | 337 | |||
| XM_017014954.1 | XP_016870443.1 | 1, 2 | 337 | |||
| GenBank mRNA | AB046844.1 | - | 1, 2 | 147 | ||
| AF376725.1 | - | 1, 2 | 157 | |||
| AK026244.1 | - | 1, 2 | 161 | |||
| AK225456.1 | - | 1, 2 | 161 | |||
| AK293092.1 | - | 1, 2 | 157 | |||
| AL834359.1 | - | 1 | 20 | |||
| BC110518.2 | - | 1, 2 | 157 | 40033360 | ||
| BC143655.1 | - | 1, 2 | 172 | |||
| BM920552.1 | - | 375 | ||||
More Information
Set Membership: |
|

GPR107
Genomic Map