siRNA Details
| Gene Symbol: EPG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020964.2 | NP_066015.2 | 27 | 4821 | ||
| XM_011526122.1 | XP_011524424.1 | 27 | 4821 | |||
| XM_017025889.1 | XP_016881378.1 | 27 | 4803 | |||
| XM_017025890.1 | XP_016881379.1 | 27 | 4803 | |||
| XM_017025891.1 | XP_016881380.1 | 26 | 4680 | |||
| XM_017025892.1 | XP_016881381.1 | 26 | 4846 | |||
| XM_017025893.1 | XP_016881382.1 | 19 | 2693 | |||
| GenBank mRNA | AK023817.1 | - | 11 | 1500 | ||
| BC130614.1 | - | 10 | 1354 | |||
| BC144104.1 | - | 10 | 1354 | |||
| BC144105.1 | - | 10 | 1354 | |||

EPG5
Genomic Map