siRNA Details
| Gene Symbol: RAB40C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172663.1 | NP_001166134.1 | 7 | 919 | ||
| NM_001172664.1 | NP_001166135.1 | 7 | 811 | |||
| NM_001172665.1 | NP_001166136.1 | 7 | 771 | |||
| NM_001172666.1 | NP_001166137.1 | 7 | 714 | |||
| NM_021168.4 | NP_066991.3 | 6 | 816 | |||
| GenBank mRNA | AY823398.1 | - | 6 | 684 | ||
| BC001581.1 | - | 3 | 410 | |||
| BC028696.1 | - | 6 | 818 | 4826800 | ||
| BX442451.2 | - | 7 | 860 | |||

RAB40C
Genomic Map