siRNA Details
| Gene Symbol: CCAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021174.5 | NP_066997.3 | 9, 10 | 1351 | ||
| NR_033902.1 | 9, 10 | 1169 | ||||
| XM_011544603.1 | XP_011542905.1 | 9, 10 | 1169 | |||
| XM_011544604.1 | XP_011542906.1 | 8, 9 | 1049 | |||
| XM_017013717.1 | XP_016869206.1 | 9, 10 | 1351 | |||
| GenBank mRNA | AK314535.1 | - | 9, 10 | 1044 | ||
| AL834162.1 | - | 9, 10 | 1338 | |||
| AL834351.1 | - | 9, 10 | 1339 | |||
| AL834352.1 | - | 9, 10 | 1339 | |||
| BX390251.1 | - | 4, 5 | 535 | |||
| BX640952.1 | - | 9, 10 | 1038 | |||

CCAR2
Genomic Map