siRNA Details
| Gene Symbol: LSM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021177.4 | NP_067000.1 | 3, 4 | 337 | ||
| GenBank mRNA | AF136977.1 | - | 3, 4 | 322 | ||
| AF182288.1 | - | 3, 4 | 156 | |||
| AF196468.1 | - | 3, 4 | 302 | |||
| AJ245416.1 | - | 3, 4 | 328 | |||
| BC009192.2 | - | 3, 4 | 319 | 3842537 | ||
| BP203102.1 | - | 3, 4 | 337 | |||
| BU189898.1 | - | 3, 4 | 328 | |||
| CR542146.1 | - | 2, 3 | 110 | |||
| CR542157.1 | - | 2, 3 | 110 | |||

LSM2
Genomic Map