siRNA Details
| Gene Symbol: LSM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021177.4 | NP_067000.1 | 5 | 857 | ||
| GenBank mRNA | AF136977.1 | - | 5 | 842 | ||
| AF182288.1 | - | 5 | 675 | |||
| AJ245416.1 | - | 5 | 848 | |||
| BC009192.2 | - | 5 | 839 | 3842537 | ||

LSM2
Genomic Map